Learn More
Diapharma Group, Inc. Anti-Hemoglobin S FITC Monoclonal Antibody, Detects HbS, Flow Cytometry Compatible, 1 mL
SDP
Supplier:  Diapharma Group, Inc. IQP574F
Encompass
The Anti-Hemoglobin S FITC Monoclonal Antibody is an Analyte-Specific Reagent (ASR) designed for detecting hemoglobin S using flow cytometry. This antibody is essential for identifying sickle cell disease through the abnormal β-chain of HbS. With a concentration of 50 µg/mL, it is compatible with various flow cytometers, providing an effective tool for research and diagnostics in hematology.
Catalog No. NC1968843