Learn More
Invivogen Anti-hCD20-Ga-hIgG1fut, Human CD20 (Obinutuzumab) antibody, non-fucosylated - Human IgG1, 100 µg
SDP
Supplier:  Invivogen HCD20GAMAB13
Encompass
100 µg purified Anti-hCD20-Ga-hIgG1fut monoclonal antibody (mAb), provided azide-free and lyophilized. Anti-hCD20-Ga-hIgG1fut is a non-fucosylated antibody featuring a human IgG1 constant region (Fc) and the variable region of Obinutuzumab (formerly named Afutuzumab or GA101) which targets the human CD20 antigen found on the surface of normal and malignant B lymphocytes.
Catalog No. NC1647735