Learn More
Immunology Consultants Monoclonal Mouse anti-hCG beta Antibody Clone#111, 1.0 mg
SDP
Supplier:  Immunology Consultants MGCB80A111
Encompass_Preferred
Monoclonal antibody specific to the beta chain of human chorionic gonadotropin.  There is no cross-reactivity with LH, TSH, or FSH, making them suitable for diagnosing early pregnancy, with a detection limit of below 10 mIU/ml.
Catalog No. NC2734269