Learn More
BIOHIPPO Anti-HCV Hepacivirin/NS3 helicase/NS3P/Viroporin p70 Antibody (28#), 100 µg
SDP
Catalog No. 5000861305
Encompass_Preferred
HCV Hepacivirin/NS3 helicase/NS3P/Viroporin p70 Recombinant Antibody (clone 28 ) is a recombinant Human IgG1, kappa antibody specific for HCV Hepacivirin/NS3 helicase/NS3P/Viroporin p70. It reacts with Hepatitis C virus genotype 3a. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61305 Supplier BIOHIPPO Supplier No. BHA21409122
HCV Hepacivirin/NS3 helicase/NS3P/Viroporin p70 Recombinant Antibody (clone 28 ) is a recombinant Human IgG1, kappa antibody specific for HCV Hepacivirin/NS3 helicase/NS3P/Viroporin p70. It reacts with Hepatitis C virus genotype 3a. Suitable for ELISA, Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.