Learn More
Immunology Consultants Monoclonal Mouse anti-Helicobacter pylori Antibody Clone# 6A4, 1.0 mg
SDP
Catalog No. NC2734286
Encompass_Preferred
The mice were inoculated with native Helicobacter Pylori extract. This mouse IgG1 is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Helicobacter Pylori lysates as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator
Catalog No. NC2734286 Supplier Immunology Consultants Supplier No. MHPL55A6A4
The mice were inoculated with native Helicobacter Pylori extract. This mouse IgG1 is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Helicobacter Pylori lysates as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator