Learn More
United States Biological Corporation ANTI-HEPARAN SULFATE DELTA
Supplier:  United States Biological Corporation H18907550UG
Encompass_Preferred
Heparan sulfate, otherwise known as heparitin sulfate or heparin monosulfate, is a generic term describing polysaccharides which are linear and consist of N-acetylated [->4)alpha-D-GlcNpAc-(1->4) �-D-GlcAp(1->] and N-sulfated disaccharides [->4)alpha-D-GlcNpS- (1->4)-�-D-GlcAp or alpha-L-IdoAp(1->] that are arranged mainly in a segregated manner. The sulfate-rich fractions in heparan sulfate are heparin-like, though they rarely possess the sulfate density found in heparin. Approximately 25% of the total polymer is initially formed by alternating arrangements of the two disaccharide units, ->4)alpha-D-GlcNpS(1->4)UAp(1->4) alpha-D-GlcNpAc(1->4)UAp (1->4)-alpha-D-GlcAp(1->. The polymer is formed as a repeating ->4)-alpha-D-GlcNpAc (1->4)-�-D-GlcAp(1-> disaccharide sequence that is attached to a serine residue of a core protein through a tetrasaccharide, glucuronosyl->galactosyl->galactosyl->xylosyl, linkage region.
Catalog No. NC0219556