Learn More
BIOHIPPO Anti-Hepatitis C virus/HCV Genome polyprotein Nanobody (SAA1346), 100 µg
SDP
Catalog No. 5000861254
Encompass_Preferred
Hepatitis C virus/HCV Genome polyprotein Nanobody Recombinant Antibody (clone SAA1346) is a recombinant Alpaca VHH-8His-Cys-tag antibody specific for Hepatitis C virus/HCV Genome polyprotein Nanobody. It reacts with Hepatitis C virus genotype 1b. Suitable for Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61254 Supplier BIOHIPPO Supplier No. BHA21407780
Hepatitis C virus/HCV Genome polyprotein Nanobody Recombinant Antibody (clone SAA1346) is a recombinant Alpaca VHH-8His-Cys-tag antibody specific for Hepatitis C virus/HCV Genome polyprotein Nanobody. It reacts with Hepatitis C virus genotype 1b. Suitable for Neutralization. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.