Learn More
Apexbio Technology LLC ANTI-HER2 PERTUZUMAB-5mg
SDP
Catalog No. 5000897593
Encompass_Preferred
Anti-ERBB2/HER2/CD340 Antibody (Pertuzumab) (CAS No 380610-27-5) is a monoclonal antibody used in cancer research particularly for targeting the ERBB2/HER2 receptor pathway which is overexpressed in certain breast cancers It functions by binding to HER2 and inhibiting dimerization preventing downstream signaling necessary for tumor growth This antibody exhibits in vitro activity in the nanomolar range effectively disrupting cell proliferation in HER2-positive cell lines Common applications include studies in cell models and animal studies to assess antitumor efficacy and it is frequently used in drug screening platforms Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97593 Supplier Apexbio Technology LLC Supplier No. F14075
Anti-ERBB2/HER2/CD340 Antibody (Pertuzumab) (CAS No 380610-27-5) is a monoclonal antibody used in cancer research particularly for targeting the ERBB2/HER2 receptor pathway which is overexpressed in certain breast cancers It functions by binding to HER2 and inhibiting dimerization preventing downstream signaling necessary for tumor growth This antibody exhibits in vitro activity in the nanomolar range effectively disrupting cell proliferation in HER2-positive cell lines Common applications include studies in cell models and animal studies to assess antitumor efficacy and it is frequently used in drug screening platforms Experimental concentrations typically depend on the specific research design