Learn More
Eagle Biosciences Inc Humanized Anti-Her2/neu (Herceptin / Trastuzumab) ELISA Assay Kit
SDP
Supplier:  Eagle Biosciences Inc AHR31K01
Encompass
The Eagle Biosciences Humanized Anti-HER-2 (Herceptin/Trastuzumab) ELISA Assay kit is intended for use in the quantitative determination of Herceptin levels in human serum and EDTA plasma. The Eagle Biosciences Humanized Anti-HER-2 (Herceptin/Trastuzumab) ELISA Assay kit is intended for research use only.
Catalog No. 50-187-8319