Learn More
Invivogen ANTI-HIFN-B-IGG 3X100UG
SDP
Supplier:  Invivogen MABG2HIFNB3
Encompass
Anti-hIFN-b-IgG -clone 10B10- 3 x 100ug. Anti-hIFN-b-IgG -clone 10B10- is a monoclonal antibody for human interferon b -hIFN-b-. This antibody has been produced in hybridomas and purified by affinity chromatography.
Catalog No. NC2310862