Learn More
Invivogen Human IL-4Rα (Dupilumab) antibody - Human IgG1, 3 x 100ug
SDP
Catalog No. NC3782333
Encompass
Anti-human IL-4R alpha antibody - Human IgG1 (high effector functions)Anti-hIL-4R-hIgG1 is a Dupilumab biosimilar with an IgG1 isotype. Dupilumab is an anti-human interleukin 4 receptor α (IL-4Rα or CD124) antibody that inhibits both IL-4 and IL-13 signaling. This monoclonal antibody (mAb) is FDA-approved for the treatment of asthma, atopic dermatitis, and chronic sinusitis.
Catalog No. NC3782333 Supplier Invivogen Supplier No. HIL4RMAB103
Anti-human IL-4R alpha antibody - Human IgG1 (high effector functions)Anti-hIL-4R-hIgG1 is a Dupilumab biosimilar with an IgG1 isotype. Dupilumab is an anti-human interleukin 4 receptor α (IL-4Rα or CD124) antibody that inhibits both IL-4 and IL-13 signaling. This monoclonal antibody (mAb) is FDA-approved for the treatment of asthma, atopic dermatitis, and chronic sinusitis.