Learn More
Genscript Corporation Antibody Resin Protein Purification Protein Purification 1 ML 1/EA
Supplier:  Genscript Corporation L004391
Encompass
GenScript Anti-His Affinity Resin is designed for purification of His-tagged protein. It is composed of agarose and conjugated THETM anti-His monoclonal antibody. His-tagged protein in the crude samples can bind to the anti-His monoclonal antibodies coupled with the agarose and the nonspecific proteins can be eliminated by washing. As a result the His-tagged protein will finally be eluted with high recovery. Moreover this product can be used for immunoprecipitation of His-tagged protein.
Catalog No. 50-252-9996