Learn More
Cayman Chemical Anti-Histone H3 pan Rabbit Mo
SDP
Catalog No. 502983195
Encompass_Preferred
Immunogen: Peptide corresponding to the C-terminus of human histone H3 • Host: Rabbit • Species Reactivity: Species independent • Cross Reactivity: (+) All histone H3 proteins independent of post-translational modifications (-) H2A, H2B, H4 • Applications: ELISA, ICC, multiplex-based assays, WB
Catalog No. 50-298-3195 Supplier Cayman Chemical Supplier No. 20724100 UG
Immunogen: Peptide corresponding to the C-terminus of human histone H3 • Host: Rabbit • Species Reactivity: Species independent • Cross Reactivity: (+) All histone H3 proteins independent of post-translational modifications (-) H2A, H2B, H4 • Applications: ELISA, ICC, multiplex-based assays, WB