Learn More
As One International Inc Anti-HIV-1 Reverse Transcriptase antibody, guinea pig serum, 50 ul
Supplier:  As One International Inc 65003
Encompass_Preferred
HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1(AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.     The product is prepared by immunizing guinea pig with HIV-1 reverse transcriptase which was uniquely over-expressed as recombinant protein in E. coli by a patent method and highly purified.     Using this antiserum in Western blotting, the bands of 66 kD and 51 kD corresponding respectively to the two subunits of HIV-1 reverse transcriptase were observed in the extract of the AIDS virus infected cells.
Catalog No. 50-165-6268