Learn More
BIOHIPPO Anti-HLA-A2-peptide (RLVDDFLLV) Complex (3H2), 100 µg
SDP
Catalog No. 5000861331
Encompass_Preferred
HLA-A2-peptide Complex (3H2) Recombinant Antibody (clone RLVDDFLLV) is a recombinant Mouse IgG1 antibody specific for HLA-A2-peptide Complex (3H2). It reacts with Human. Suitable for ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-61331 Supplier BIOHIPPO Supplier No. BHA21409348
HLA-A2-peptide Complex (3H2) Recombinant Antibody (clone RLVDDFLLV) is a recombinant Mouse IgG1 antibody specific for HLA-A2-peptide Complex (3H2). It reacts with Human. Suitable for ELISA. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.