Learn More
CAPRICO BIOTECHNOLOGIES INC Caprico Bio, Fluorescent Antibody Conjugates
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4032025
Encompass_Preferred
Caprico Bio's fluorescent antibody conjugation kits enable quick and efficient labeling of antibodies with various bright fluorophores like Alexa Fluor, CF, and DyLight dyes. These kits, complete with all necessary reagents, facilitate antibody conjugation in 30-60 minutes for applications such as flow cytometry, cell imaging, ELISA, and Western blot. They are designed for high efficiency and maintain antibody functionality, offering a range of dyes for multicolor detection in research.
Catalog No. 50-227-9288