Learn More
Bio X Cell Antibody, Human MHC, Clone W6/32, 5 MG
Supplier: Bio X Cell BE0079R005MG
The W6/32 monoclonal antibody reacts with human major histocompatibility complex (MHC) class I, HLA-A, B, and C. All human nucleated cells express MHC class I antigens associated with β2-microglobulin. MHC class I plays a central role in cell-mediated immune responses and tumor surveillance.
- Clone W6/32
- Reacts with human MHC Class I (HLA-A, HLA-B, HLA-C)
- Isotype mouse IgG2a, κ
- Unconjugated
- Purity ≥95%
- Endotoxin ≤1EU/mg
- Sterility achieved via 0.2 μm filtration
- Store at 4°C, do not freeze
- Suitable for functional assays, in vitro organoids/organ-on-chip, immunopeptidomics, immunoprecipitation, immunofluorescence, flow cytometry, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.