Learn More
Sino Biological Anti-HMGB1 Antibody, Mouse Monoclonal 100 µL
Catalog No. 5000749872
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HMGB1 (rh HMGB1; Catalog#10326-H07E; P09429; Lys57-Asp158). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-49872 Supplier Sino Biological Supplier No. 10326MM22100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HMGB1 (rh HMGB1; Catalog#10326-H07E; P09429; Lys57-Asp158). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.