Learn More
Biolegend Purified anti-human CD19 (Maxpar Ready)
Supplier:  Biolegend 302247
Encompass_Preferred
Purified anti-human CD19 (Maxpar Ready) [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC, CyTOF; Size: 100 ug
Catalog No. NC1476462