Learn More
Seracare Life Sciences Inc ANTI-HORSE IGG (G) AB FITC
Supplier:  Seracare Life Sciences Inc 52300318
Encompass_Preferred
Affinity purified polyclonal antibody to Horse IgG, heavy chains (gamma) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-669-04