Learn More
Seracare Life Sciences Inc ANTI-HORSE IGG (H+L) AB BIOTIN
Supplier:  Seracare Life Sciences Inc 52600056
Encompass_Preferred
Affinity purified polyclonal antibody to Horse IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-85