Learn More
Immunology Consultants anti-Horse SAA Antibody (Mouse - Clone #4B2) - Monoclonal
SDP
Catalog No. 5000435479
Encompass_Preferred
This monoclonal antibody provides high‑specificity detection of equine Serum Amyloid A, a key biomarker for inflammation and systemic disease in horses. Clone 4B2 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays. MSAA‑70A‑4B2 is ideal for equine diagnostics, performance‑health monitoring, and research on acute‑phase responses. Its consistent performance ensures reliable detection across serum, plasma, and purified antigen preparations
Catalog No. 50-004-35479 Supplier Immunology Consultants Supplier No. MSAA70A4B2
This monoclonal antibody provides high‑specificity detection of equine Serum Amyloid A, a key biomarker for inflammation and systemic disease in horses. Clone 4B2 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays. MSAA‑70A‑4B2 is ideal for equine diagnostics, performance‑health monitoring, and research on acute‑phase responses. Its consistent performance ensures reliable detection across serum, plasma, and purified antigen preparations