Learn More
Immunology Consultants anti-Horse SAA Antibody (Mouse - Clone #4F6) - Monoclonal
SDP
Catalog No. 5000435398
Encompass_Preferred
Clone 4F6 targets a distinct equine SAA epitope, providing complementary detection to Clone 4B2. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MSAA‑70A‑4F6 is well suited for veterinary diagnostics, inflammation research, and monitoring equine acute‑phase responses. Its reproducible performance ensures dependable detection across repeated assays.
Catalog No. 50-004-35398 Supplier Immunology Consultants Supplier No. MSAA70A4F6
Clone 4F6 targets a distinct equine SAA epitope, providing complementary detection to Clone 4B2. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MSAA‑70A‑4F6 is well suited for veterinary diagnostics, inflammation research, and monitoring equine acute‑phase responses. Its reproducible performance ensures dependable detection across repeated assays.