Learn More
Charles River Labs Cell Solutions Anti-HPV E7 (11-19) CD8+ T Cells, 1 to 2 Million cells/vial, Cryopreserved
Supplier:  Charles River Labs Cell Solutions ASTC1099
Encompass
These are CD8 T cells specific for the E7 protein of human papilloma virus. The antigenic peptide sequence is YMLDLQPET representing residues 11-19 of the protein and it is presented by the HLA-A 02:01 allele.
Catalog No. 50-225-6108
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov