Learn More
Enzo Life Sciences Anti-HSP70 IgG/A/M (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS750
Encompass_Preferred
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available. This assay allows for reproducible, accurate and precise determination of IgG , IgA and IgM antibodies (total) to human Hsp70 in serum. The ELISA kit uses recombinant human Hsp70 bound to the wells of the plate to bind anti-human Hsp60 antibodies present in human serum. The captured anti-human Hsp70 antibodies are detected with a HRP conjugated goat polyclonal antibody specific for human IgG, IgA and IgM molecules. The assay is developed with tetramethylbenzidine substrate pro ducing a blue color in proportion to the amount of captured anti -human Hsp70 antibodies. Alternative name: Heat shock protein 70, HSP72. Sensitivity: 6.79 ng/ml (range 31.25 - 1,000 ng/ml). Assay Time: <4 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human . Content: Microtiter plate, Standard, Sample diluent 2, Wash buffer concentrate, HRP Conjugate, TMB Substrate, Stop solution 2.
Catalog No. 50-200-7218