Learn More
Seracare Life Sciences Inc ANTI-HU A+G+M (H+L) MSA FITC
Supplier:  Seracare Life Sciences Inc 52300290
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Human IgA, IgG, and IgM, both heavy and light chains (whole IgA, IgM, and IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC).
Catalog No. 50-668-76