Learn More
BIOGEMS INTERNATIONAL INC Anti-Human CD11b PE-Cy7 Size 100 tests
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 0321177100
Encompass
The ICRF44 monoclonal antibody specifically reacts with the 165 kDa human adhesion glycoprotein CD11b, which forms, together with the 95 kDa CD18 (integrin β2) a complex known as Mac-1. CD11b is expressed on the surface of activated lymphocytes, a subset of natural killer cells, granulocytes, and monocytes. It functions as a receptor in cell-cell and cell-matrix interactions and as a receptor for iC3b, ICAM-1, ICAM-2, and ICAM-3 intercellular adhesion molecules.
Catalog No. 50-152-8760