Learn More
Leinco Technologies Inc Anti-Human CD14 (P35, LPS receptor) In Vivo GOLD; Clone UCHM-1; Isotype: Mouse IgG2a
SDP
Supplier:  Leinco Technologies Inc C37025 MG
Encompass_Preferred
Specificity: Clone UCHM-1 recognizes Human CD14. Antigen Distribution: CD14 is expressed on monocytes, macrophages, dendritic cells, and granulocytes. Background: CD14 is a 55 kDa GPI-anchored single chain glycoprotein that enhances activation of cells by lipopolysaccharide (LPS). CD14 is a high-affinity receptor for LPS. As such, it is involved in the upregulation of adhesion molecules and cytokine expression in monocytes and neutrophils. CD14 also plays a role in the clearance of gram-negative pathogens. Applications: (FC, WB) Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6078