Learn More
Fluidigm Corp ANTI-HU CD15 -164DY
SDP
Supplier:  Fluidigm Corp 3164001B
Encompass
CD15, also known as LewisX, 3-FAL and SSEA-1, is a carbohydrate adhesion molecule, 3-fucosyl-N-acetyllactosamine (3-FAL). It is expressed by granulocytes, in varying degrees by monocytes, and in patients with Hodgkin's disease, some B cell chronic lymphocytic leukemias, acute lymphoblastic leukemias, and most acute nonlymphocytic leukemias.
Catalog No. 50-466-926