Learn More
Biolegend Alexa Fluor 700 anti-human CD161
Supplier:  Biolegend 339941
Encompass_Preferred
Alexa Fluor 700 anti-human CD161 [HP-3G10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. NC1923570