Learn More
Fluidigm Corp ANTI-HU CD183 CXCR3 -156GD
SDP
Supplier:  Fluidigm Corp 3156004B
Encompass
CXCR3, also known as G protein-coupled receptor 9 (GPR9) and CD183, is a 7-pass transmembrane receptor coupled to G protein. It is a chemokine receptor that binds CXCL9, CXCL10, and CXCL11. CXCR3 is highly expressed by T lymphocytes, NK cells, dendritic cells, mast cells, alveolar macrophages, eosinophils, and human airway epithelial cells.
Catalog No. 50-466-886