Learn More
Fluidigm Corp ANTI-HU CD3 -170ER
SDP
Supplier:  Fluidigm Corp 3170001B
Encompass
CD3 is a transmembrane subunit of the T cell receptor (TCR) complex. It is a member of the immunoglobulin superfamily that plays a role in antigen recognition, signal transduction, and T cell activation. CD3 is expressed by T lymphocytes (thymocytes) at the highest levels on mature cell types.
Catalog No. 50-466-972