Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Monoclonal Anti-Human CD3 Antibody (Clone OKT3), Mouse IgG2a, Unconjugated, 1 mg

Catalog No. NC3782624
Encompass_Preferred

Monoclonal anti-human CD3 antibody (clone OKT3) is a recombinant mouse IgG2a, kappa antibody expressed in CHO cells and supplied unconjugated as a 1 mg vial for preclinical research. It binds the CD3ε component of the human T-cell receptor complex and is suitable for functional cell-based assays, including stimulation of IL-2 secretion (QC tested EC50 ~0.15 ng/mL).

  • Clone: OKT3
  • Isotype: Mouse IgG2a, kappa
  • Host: Mouse
  • Expression system: CHO cells
  • Format: Unconjugated antibody
  • Size: 1 mg
  • Functional activity: Stimulates IL-2 secretion; EC50 0.15 ng/mL (QC tested)
  • Applications: Cell culture and early preclinical assays

Catalog No. NC3782624 Supplier ACROBiosystems Supplier No. CDECH44G11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoclonal anti-human CD3 antibody (clone OKT3) is a recombinant mouse IgG2a, kappa antibody expressed in CHO cells and supplied unconjugated as a 1 mg vial for preclinical research. It binds the CD3ε component of the human T-cell receptor complex and is suitable for functional cell-based assays, including stimulation of IL-2 secretion (QC tested EC50 ~0.15 ng/mL).

  • Clone: OKT3
  • Isotype: Mouse IgG2a, kappa
  • Host: Mouse
  • Expression system: CHO cells
  • Format: Unconjugated antibody
  • Size: 1 mg
  • Functional activity: Stimulates IL-2 secretion; EC50 0.15 ng/mL (QC tested)
  • Applications: Cell culture and early preclinical assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.