Learn More
Leinco Technologies Inc Anti-Human CD71 (Transferrin Receptor, TFR, T9) In Vivo PLATINUM; Clone T56/14; Isotype: Mouse IgG1
SDP
Supplier:  Leinco Technologies Inc C6711.0 MG
Encompass_Preferred
Specificity: Clone T56/14 recognizes Human CD71. Antigen Distribution: CD71 is expressed on most proliferating cells, activated lymphocytes, monocytes, macrophages, erythroid progenitors, and brain endothelium. Background: CD71 is a 95 kDa type II transmembrane glycoprotein that forms a disulfide-linked homodimer (Mr 190 kDa). CD71 is involved in mediating cellular proliferation by facilitating the uptake of iron via ferrotransferrin binding and the recycling of apotransferrin to the cell surface. Applications: (FC, WB) Clone T56/14 may be used to identify activated lymphocytes. It may also be used in studies of erythropoiesis. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6213