Learn More
Leinco Technologies Inc Anti-Human EGFR (Epidermal Growth Factor Receptor, ERBB, ERBB1, mENA) In Vivo PLATINUM; Clone EGFR.1; Isotype: Mouse IgG2b κ
SDP
Supplier:  Leinco Technologies Inc E33725 MG
Encompass_Preferred
Specificity: Clone EGFR.1 recognizes the human EGFR. Antigen Distribution: EGFR is found in epithelial cells (including epithelial carcinomas) and endothelial cells. Background: EGFR is a 170kDa transmembrane glycoprotein that is part of the protein kinase superfamily and controls cell growth and differentiation. Ligand binding induces dimerization and autophosphorylation. It consists of a glycosylated extracellular domain which binds to EGF and an intracellular domain with tyrosine-kinase activity necessary for signal transduction. TGFα, vaccinia virus growth factor, and related growth factors can also bind to and signal through EGFR. Applications: (WB) Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6286