Learn More
Leinco Technologies Inc Anti-Human HLA-A, B, C (MHC class I) In Vivo GOLD; Clone W6/32; Isotype: Mouse IgG2a
SDP
Supplier:  Leinco Technologies Inc H2631.0 MG
Encompass_Preferred
Specificity: Clone W6/32 recognizes human major histocompatibility complex (MHC) class I, HLA-A, B, and C. Antigen Distribution: MHC class I antigens associated with β2-microglobulin are expressed by all human nucleated cells. HLA-A, B, C is present on most nucleated cells of humans and old world monkeys. Background: MHC class I molecules are part of the Ig superfamily and are involved in the presentation of antigens to CD8+ T cells. HLA-A, B, C plays a central role in cell-mediated immune responses and tumor surveillance. Applications: (WB) Clone W6/32 recognizes residues in the N terminus of the human β2-microglobulin molecule. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6345