Learn More
Seracare Life Sciences Inc ANTI-HU IGA (ALPHA) AB FITC
Catalog No. 5066870
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgA heavy chains (alpha) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-668-70 Supplier Seracare Life Sciences Inc Supplier No. 52300284
Affinity purified polyclonal antibody to Human IgA heavy chains (alpha) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.