Learn More
Seracare Life Sciences Inc ANTI-HU IGA+IGG+IGM (H+L) AB
Catalog No. 5066807
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Human IgA, human IgG, and human IgM, both heavy and light chains (whole IgA, IgM, and IgG) made in Goat. Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-668-07 Supplier Seracare Life Sciences Inc Supplier No. 52100160
Affinity purified polyclonal secondary antibody against Human IgA, human IgG, and human IgM, both heavy and light chains (whole IgA, IgM, and IgG) made in Goat. Product is in lyophilized form.