Learn More
Seracare Life Sciences Inc ANTI-HU IGA+IGG+IGM(H+L)AB MSA
Supplier:  Seracare Life Sciences Inc 52100164
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Human IgA, IgG, and IgM, both heavy and light chains (whole IgA, IgM, and IgG) made in Goat. Product has been cross-adsorbed with mouse immunoglobulin to reduce cross-reactivity.
Catalog No. 50-668-08