Learn More
Seracare Life Sciences Inc ANTI-HU IGE (E) AB BIOTIN
Supplier:  Seracare Life Sciences Inc 52600030
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgE heavy chains (epsilon) made in Goat and biotinylated. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-58