Learn More
Seracare Life Sciences Inc ANTI-HU IGE (EPSILON)AB FITC
Catalog No. 5066873
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgE heavy chains (epsilon) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-668-73 Supplier Seracare Life Sciences Inc Supplier No. 52300287
Affinity purified polyclonal antibody to Human IgE heavy chains (epsilon) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.