Learn More
Seracare Life Sciences Inc ANTI-HU IGE (EPSILON) AB HRP
Supplier:  Seracare Life Sciences Inc 52200329
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgE heavy chains (epsilon) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-671-94