Learn More
Seracare Life Sciences Inc ANTI-HU IGG (G) AB RESERVEAP
Catalog No. 5067393
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with alkaline phosphatase. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-673-93 Supplier Seracare Life Sciences Inc Supplier No. 52200462
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with alkaline phosphatase. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.