Learn More
Seracare Life Sciences Inc ANTI-HU IGG (GAMMA) AB FITC
Catalog No. 5066871
Encompass_Preferred
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-668-71 Supplier Seracare Life Sciences Inc Supplier No. 52300285
Affinity purified polyclonal antibody to Human IgG heavy chains (gamma) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.