Learn More
Seracare Life Sciences Inc ANTI-HU IGG (H+L) F(AB )2 FITC
Supplier:  Seracare Life Sciences Inc 52300443
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Human IgG, heavy and light chains (whole IgG). Made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-54