Learn More
Seracare Life Sciences Inc ANTI-HU IGG (H+L) F(AB )2 HRP
Catalog No. 5067363
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Human IgG, heavy and light chains (whole IgG). Made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-673-63 Supplier Seracare Life Sciences Inc Supplier No. 52200392
Affinity purified polyclonal antibody, F(ab)2 fragment, to Human IgG, heavy and light chains (whole IgG). Made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.