Learn More
Leinco Technologies Inc Anti-Human TCR Vα24-Jα18 (TCR Vα24-JαQ, invariant NKT cell, iNKT cell) In Vivo PLATINUM; Clone 6B11; Isotype: Mouse IgG1 κ
SDP
Supplier:  Leinco Technologies Inc T17055.0 MG
Encompass_Preferred
Specificity: Clone 6B11 recognizes TCR Vα24-Jα18. Antigen Distribution: TCR Vα24-Jα18 is expressed primarily on invariant NKT cells, which are a small subset of T lymphocytes. Background: Invariant NKT cells have been reported to play a role in immune regulation, tumor surveillance, and host response to pathogens. They can be specifically activated by CD1d-presented antigen resulting in immediate production of IL-4 and IFNγ. Applications: (WB) Clone 6B11 reacts with a unique epitope in the CDR3 region of TCR Vα24-Jα18 and is able to identify a subset of all Vα24 positive T cells. The binding of the PE-conjugate of 6B11 may be partly blocked by antibodies specific for Vα24. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6641