Learn More
Echelon Biosciences Research Labs Anti-hVps34 Antibody for IP 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZR01550UG
Encompass
Z-R015 is an affinity purified rabbit polyclonal anti-hVps34 antibody. It can immunoprecipicate human Vps34 from cell lysate without inhibition. There is no need to elute the enzyme prior to running and activity assay. See figure above for hVps34 activity data obtained using Z-R015 in IP. This item is non returnable
Catalog No. 50-720-926