Learn More
Echelon Biosciences Research Labs Anti human Vps34 Antibody 10 ug
SDP
Supplier:  Echelon Biosciences Research Labs ZR01610UG
Encompass
Z-R016 is an affinity purified rabbit polyclonal anti human Vps34 antibody. The main application for this is as a primary antibody for Western Blot at 2.5 g/mL concentration. Z-R016 works well with immunoprecipitated hVps34 (Fig.1 above) or partially purified hVps34. The anti human Vps34 antibody has also been validated for use with whole cell lysate (refer to Fig.2 in TDS file) and immunostaining This item is non returnable
Catalog No. 50-720-927