Learn More
Beckman Coulter CD335-PE, BAB281, 1 mL, ASR
Supplier:  Beckman Coulter IM3711
Encompass
CD335, also known as NKp46, is a 46-kDa transmembrane glycoprotein which is a NK-specific triggering receptor involved in non-MHC-restricted natural cytotoxicity and considered the prototype of the NK natural cytotoxicity receptors (NCRs).
Catalog No. NC0627958